Cuvettes for fluorescence
measurements
Supplier: Agilent Technologies
VARI6610023700EA 1880 EUR
VARI6610023700 VARI6610021400 VARI6610000900 VARI6610021500 VARI6610023500 VARI6610021200 VARI6610021300 VARI6610024200 VARI6610001200 VARI6610021600
Cuvettes for fluorescence
measurements
Cuvettes Spectrophotometer Cuvettes
These cuvettes are designed for use with Cary Eclipse fluorescence spectrophotometer.
Applications include academia, Life sciences, biotechnology, chemicals, energy and food analyses.