Tubing
Supplier: Agilent Technologies
VARICP4260EA 33.3 EUR
VARICP4260 VARIR000383005 HEWL5190-6983 VARICP4008 HEWL5062-2419 VARIAL3003 VARIAL3002 VARIAL3000
Tubing
Tubing Chromatography Tubing
Copper and stainless steel tubing for gas chromatography (GC) applications.
  • Copper can be used for most applications, except those requiring the highest purity gas delivery system
  • Stainless steel and UltiMetal Plus tubing are appropriate for more demanding applications
These high-quality copper, stainless steel, and UltiMetal Plus tubing are manufactured and cleaned for chromatographic use.