PEEK tubing
Supplier: Agilent Technologies
VARIAL35722EA 0 EUR
VARIAL35722 VARIAL35723 VARIAL35702 VARIAL35724 VARIAL35725 VARIAL35703 VARIAL35726 VARIAL35705 VARIAL35727 VARIAL35729 VARIAL35707 VARIAL35741 VARIAL35720 VARIAL35708 VARIAL35709 VARIAL35711 VARIAL35712 VARIAL35713 VARIAL35714 VARIAL35715 VARIAL35716 VARIAL35717 VARIAL35718 VARIAL35710 VARIAL35719
PEEK tubing
Tubing Chromatography Tubing
Polymer tubing
  • For high-pressure HPLC plumbing
  • Maximum temperature: 100 °C