Retention gaps, fused silica and UltiMetal™
Supplier: Agilent Technologies
VARICP8068EA 0 EUR
VARICP8068 VARICP8089 VARICP8015 VARICP8017 VARICP8008 VARICP8019 VARICP8018 VARICP8007 VARICP8070 VARICP8066 VARICP8088 VARICP8087 VARICP8080 VARICP8090 VARICP8009
Retention gaps, fused silica and UltiMetal™
Tubing Peristaltic Pump Tubing
The fused silica retention gaps has three different polarities, uncoated for universal applicability. Dimensions equal to 0,53 mm int. Ø fused silica provide easy installation.
  • Easy to install
  • Single continuous length with leak free connections
These fused silica retention gaps with universal CP-Quick-Seal connectors that fit all capillary column sizes. UltiMetal™ retention gaps are complete with coupled high temperature glass-lined connector for ease-of-use.