Stereo microscopes, ergonomic, Mantis
Supplier: Vision Engineering
VISIMEO-006EA 559 EUR
VISIMEO-006 VISIMC-UNI VISIMEO-015 VISIMEO-004 VISIMEO-002 VISIMEO-010 VISIMC-ST VISIME-ST VISIMECAM-ST VISIMEO-008 VISIMCO-008 VISIMCO-006 VISIMEO-020 VISIMECAM-UNI VISIMCO-004 VISIMCO-002 VISIME-UNI
Stereo microscopes, ergonomic, Mantis
Microscopes
Superior imaging for a wide range of inspection and rework tasks.
  • Patented optical technology for fatigue-free viewing and superb image quality
  • Wide range of magnification options to 20x
  • Long working distances; large depths of field
  • Shadow-free true colour LED illumination
  • Choice of table stand or articulated arm
Mantis Compact excels in the low magnification range for inspection or manipulation tasks where bench magnifiers have traditionally been used. Mantis Compact has a small footprint and low nvestment cost giving a superb price/performance ratio.
Mantis Elite has enhanced optical performance, including higher magnification, a large field of view plus long working distances, making it a perfect alternative to traditional stereo microscopes for a wide range of inspection, preparation and manipulation tasks requiring hand-eye co-ordination.
Ordering information: Objective lenses sold separately.