Reciprocating shaker, SHR digital
Supplier: Witeg
WITGDH.WSR04022EA 927.05 EUR
WITGDH.WSR04022 WITGDH.WSR04020
Reciprocating shaker, SHR digital
Shakers Reciprocating Shakers
These digital reciprocating shakers with programmable digital feedback control system are ideal for culturing cells in hybridisation, staining and destaining gels. Also used in combinatorial chemistry.
  • Brushless DC motor (low noise and long life cycle)
  • High precision digital rotation speed control
  • Over-Load protection, storage function and alarm
  • Programmable digital feedback controller with Jog-Shuttle switch (turn + push)
The optimal platform rotation helps for intensive mixing with convenient setting of timer and speed using jog shuttle switch. For experimental safety shakers are supported with locking mode in which input to jog-shuttle can be disabled and it also contain programmable operation modes.
Ordering information: Flask holder, rubber mat or universal platform for easy and fast fixation for any kind of flasks and bottles are available on request.