Cyclosporin A

Supplier: Thermo Fisher Scientific
J63191.03EA 207 EUR
J63191.03 J63191.06
Cyclosporin A
Proteins and Peptides
White to off-white crystalline solid

  • Soluble in DMSO or ethanol. Insoluble in water.

Signal word: Danger
GHS Symbols: GHS08, GHS07
Purity: 99+%
CAS No.: 59865-13-3
Molecular Formula: C₆₂H₁₁₁N₁₁₀₁₂
Formula Weight: 1202.61
Storage Temperature: –30 to –10°C
Physical Form: Crystalline solid
Melting Point: 146 to 151°
Solubility: Soluble in DMSO or ethanol. Insoluble in water.
Beilstein Reference: 3647785
Merck Reference: 14,2752
MDL No.: MFCD00274558

Immunosuppressant. Induces apoptosis and inhibits angiogenesis induced by VEGF.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR