Hydrofluoric acid 4,9%, CMOS for microelectronic, J.T.Baker®
|
|
5397-03EA
0
EUR
5397-03
Hydrofluoric acid 4,9%, CMOS for microelectronic, J.T.Baker®
Hydrofluoric acid
Formula:
HF MW: 20,01 g/mol |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 EINECS: 231-634-8 UN: 1790 ADR: 8,II |
Specification Test Results
For Microelectronic Use | |
Assay (HF) | 4.80 - 5.00 % |
Color (APHA) | ≤10 |
Fluosilicic Acid (H₂SiF₆) | ≤0.010 % |
Residue after Ignition | ≤1 ppm |
Chloride (Cl) | ≤3 ppm |
Nitrate (NO₃) | ≤2 ppm |
Phosphate (PO₄) | ≤0.7 ppm |
Sulfate and Sulfite (as SO₄) | ≤3 ppm |
Trace Impurities - Aluminum (Al) | ≤50.0 ppb |
Arsenic and Antimony (as As) | ≤30.0 ppb |
Trace Impurities - Barium (Ba) | ≤20.0 ppb |
Trace Impurities - Boron (B) | ≤20.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤20.0 ppb |
Trace Impurities - Calcium (Ca) | ≤50.0 ppb |
Trace Impurities - Chromium (Cr) | ≤10.0 ppb |
Trace Impurities - Cobalt (Co) | ≤20 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤20.0 ppb |
Trace Impurities - Germanium (Ge) | ≤50.0 ppb |
Trace Impurities - Gold (Au) | ≤10.0 ppb |
Heavy Metals (as Pb) | ≤100 ppb |
Trace Impurities - Iron (Fe) | ≤50.0 ppb |
Trace Impurities - Lead (Pb) | ≤20.0 ppb |
Trace Impurities - Lithium (Li) | ≤20.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤50.0 ppb |
Trace Impurities - Manganese (Mn) | ≤20.0 ppb |
Trace Impurities - Nickel (Ni) | ≤20.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - Silicon (Si) | ≤1000.0 ppb |
Trace Impurities - Silver (Ag) | ≤10.0 ppb |
Trace Impurities - Sodium (Na) | ≤100.0 ppb |
Trace Impurities - Strontium (Sr) | ≤20.0 ppb |
Trace Impurities - Tin (Sn) | ≤50 ppb |
Trace Impurities - Titanium (Ti) | ≤50.0 ppb |
Trace Impurities - Zinc (Zn) | ≤50 ppb |
Particle Count - 0.5 µm and greater | ≤60 par/ml |
Particle Count - 1.0 µm and greater | ≤8 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...