Aluminium chloride 28% in aqueous solution, Sigma-Aldrich®

SIAL11018-1000KG-HEA 3530 EUR
HONE11018.91000 HONE11018.9070 SIAL11018-1000KG-H
Aluminium chloride 28% in aqueous solution, Sigma-Aldrich®
Aluminium chloride
Formula: AlCl₃
MW: 133,34 g/mol
MDL Number: MFCD00003422
UN: 2581
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR