2-(Hydroxymethyl)-1,4-benzodioxane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIALH38403-5GEA
46.5
EUR
SIALH38403-5G
2-(Hydroxymethyl)-1,4-benzodioxane, Sigma-Aldrich®
2-(Hydroxymethyl)-1,4-benzodioxane
Formula:
C₉H₁₀O₃ MW: 166,18 g/mol |
MDL Number:
MFCD00006822 CAS Number: 3663-82-9 EINECS: 222-921-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...