Headspace crimp top vials and closures, 20 mm
Supplier: Thermo Fisher Scientific
HYPE60180-504EA 103 EUR
HYPE60180-504 HYPE60180-505
Headspace crimp top vials and closures, 20 mm
Vials
Clear glass vials with 20 mm crimp seal finish are designed to fit most headspace autosamplers.
  • Made of superior quality 1st hydrolytic class glass (Type 1) in clear and amber
Certifications: Glass meets all requirements of Pharm. US, EU, JPN.