Sample dispensing systems, eVol™ XR
Supplier: Thermo Scientific
eVol®
HYPE66002-022EA 115 EUR
HYPE66002-022 HYPE66002-020 HYPE66002-021 HYPE66002-023
Sample dispensing systems, eVol™ XR
Syringes Syringes for Chromatography
The eVol™ XR dispensing system is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures.
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full colour screen
The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing.
Certifications: Complies with GLP and GMP guidelines.
Ordering information: The kit includes an eVol™ XR dispensing unit, three syringes (5, 50, and 500 µl), charger, stand and manual. The stand-alone unit does not contain charger or other accessories, these have to be ordered separately.