These all-in-one systems with guided menu navigation make general water quality meter testing fast and simple.
- Simple and fast measurements
- Measure electrical conductivity and salinity (NaCl), the GLP meter (EC71) also measures total dissolved solids (TDS)
- Data Logger for 400 measurements, calibration data and cell record (Only GLP meter EC71)
- Data can be sent to printer or PC through the RS 232 C interface (Only GLP meter EC71)
Each system is designed to be used in a wide variety of applications and comes complete with everything you need to start testing. A guided navigation menu allows you to follow simple prompts to set up your measurements thus reducing the time necessary to start testing, and improving the accuracy of measurements.
Delivery information: The sensION+ EC7 benchtop conductivity kit is supplied with integrated magnetic stirrer and probe holder, power supply with interchangeable plugs, sensION+ 5070 conductivity cell with temperature sensor, 147 µS/cm, 1413 µS/cm, 12,88 mS/cm conductivity standard solutions, 3 calibration beakers and printed documentation.
The sensION+ EC71 (GLP) meter is supplied with integrated magnetic stirrer and probe holder, power supply with interchangeable plugs, 147 µS/cm, 1413 µS/cm, 12,88 mS/cm conductivity standard solutions, 3 calibration beakers and printed documentation.
The EC71 kit additionally includes the sensION+ 5070 conductivity cell with temperature sensor.