Anti-GFAP Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAM1870BEA 348 EUR
ABGEAM1870B
Anti-GFAP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR