Anti-CHPT1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP10186AEA 348 EUR
ABGEAP10186A
Anti-CHPT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CHPT1 (choline phosphotransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR