Anti-MRPS12 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP12638CEA 348 EUR
ABGEAP12638C
Anti-MRPS12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MRPS12 (mitochondrial ribosomal protein S12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR