Anti-PGAP3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP13237CEA 348 EUR
ABGEAP13237C
Anti-PGAP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PGAP3 (post-GPI attachment to proteins 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR