Anti-MAGEA1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP6163AEA 360 EUR
ABGEAP6163A
Anti-MAGEA1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAGEA1 (melanoma antigen family A, 1 (directs expression of antigen MZ2-E))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR