Anti-ST13 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP6247AEA 348 EUR
ABGEAP6247A
Anti-ST13 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ST13 (suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR