Anti-SNCA Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP6401BEA 348 EUR
ABGEAP6401B
Anti-SNCA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SNCA (synuclein, alpha (non A4 component of amyloid precursor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR