Anti-Sestrin-1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP7650BEA 350 EUR
ABGEAP7650B
Anti-Sestrin-1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Sestrin-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR