Anti-MSH2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNB100-56428EA 425 EUR
NOVUNB100-56428
Anti-MSH2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MSH2 (mutS homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR