Anti-Oval Cell Marker Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: OC2-1D11]

Supplier: Novus Biologicals
NOVUNBP1-18963APCEA 656 EUR
NOVUNBP1-18963APC
Anti-Oval Cell Marker Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: OC2-1D11]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-1D11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR