Anti-Oval Cell Marker Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: OC2-2F3]

Supplier: Novus Biologicals
NOVUNBP1-18964PEEA 656 EUR
NOVUNBP1-18964PE
Anti-Oval Cell Marker Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: OC2-2F3]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-2F3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR