Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 550) [clone: OC2-2A6]

Supplier: Novus Biologicals
NOVUNBP1-18967REA 656 EUR
NOVUNBP1-18967R
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 550) [clone: OC2-2A6]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-2A6
Conjugation: DyLight® 550
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR