Anti-Clara Cell Protein Mouse Monoclonal Antibody [clone: 13B183]

Supplier: United States Biological
USBI030105EA 590 EUR
USBI030105
Anti-Clara Cell Protein Mouse Monoclonal Antibody [clone: 13B183]
Antibodies
Type: Primary
Antigen: Clara Cell Protein
Clonality: Monoclonal
Clone: 13B183
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR