Anti-SNX30 Rabbit Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBI042122-BIOTINEA 852 EUR
USBI042122-BIOTIN
Anti-SNX30 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SNX30 (sorting nexin family member 30)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR