Anti-ZNF816-ZNF321P Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: United States Biological
USBI044188-FITCEA 852 EUR
USBI044188-FITC
Anti-ZNF816-ZNF321P Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: ZNF816-ZNF321P (ZNF816-ZNF321P readthrough)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR