Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 10G12]

Supplier: US Biological
USBI125915EA 522 EUR
USBI125915
Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 10G12]
Antibodies
Recommended dilution: Immunochemistry: 1. 2?g/ml

Type: Primary
Antigen: DNA Ligase 1
Clonality: monoclonal
Clone: 10G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR