Anti-HLCS Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI127936EA 561 EUR
USBI127936
Anti-HLCS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HLCS (holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase (ATP-hydrolysing)) ligase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR