Anti-Mesenchymal Stem Cell Protein DSCD75 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI136252EA 633 EUR
USBI136252
Anti-Mesenchymal Stem Cell Protein DSCD75 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 7. 5-10 ?g/ml

Type: Primary
Antigen: Mesenchymal Stem Cell Protein DSCD75
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR