Anti-Mast Cell Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI138118EA 1250 EUR
USBI138118
Anti-Mast Cell Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Mast Cell
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR