Anti-GJA5 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI144383EA 554 EUR
USBI144383
Anti-GJA5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GJA5 (gap junction protein, alpha 5, 40kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR