Anti-Platelet endothelial cell adhesion molecule Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI145063EA 516 EUR
USBI145063
Anti-Platelet endothelial cell adhesion molecule Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Platelet endothelial cell adhesion molecule
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR