Anti-Biotin Ligase Epitope Tag Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI168059EA 714 EUR
USBI168059
Anti-Biotin Ligase Epitope Tag Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:5000; Western Blot: 1:5000

Type: Tag
Antigen: Biotin Ligase Epitope Tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR