Anti-B Cell Subset Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K110]

Supplier: United States Biological
USBIB0001-65CEA 446 EUR
USBIB0001-65C
Anti-B Cell Subset Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K110]
Antibodies
Type: Primary
Antigen: B Cell Subset
Clonality: monoclonal
Clone: 5K110
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR