Anti-B Cell Subset Mouse Monoclonal Antibody [clone: 10B247]

Supplier: United States Biological
USBIB0001-65DEA 836 EUR
USBIB0001-65D
Anti-B Cell Subset Mouse Monoclonal Antibody [clone: 10B247]
Antibodies
Recommended dilution: Flow Cytometry:

Type: Primary
Antigen: B Cell Subset
Clonality: monoclonal
Clone: 10B247
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR