Anti-Cytokeratin, Basal Cell Mouse monoclonal antibody [clone: RCK]

Supplier: United States Biological
USBIC9097-03CEA 699 EUR
USBIC9097-03C
Anti-Cytokeratin, Basal Cell Mouse monoclonal antibody [clone: RCK]
Antibodies
Recommended dilution: Western Blot: 1:10-1:25; Immunochemistry: 1:10-1:25;

Type: Primary
Antigen: Cytokeratin, Basal Cell
Clonality: Monoclonal
Clone: RCK
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Dog, Guinea pig, Hamster, Human, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR