Anti-Defender of Cell Death 1 Rabbit polyclonal antibody

Supplier: United States Biological
USBID1877-15EA 526 EUR
USBID1877-15
Anti-Defender of Cell Death 1 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: Defender of Cell Death 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR