Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 10H5]

Supplier: United States Biological
USBID3925-01LEA 633 EUR
USBID3925-01L
Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 10H5]
Antibodies
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DNA Ligase 1
Clonality: monoclonal
Clone: 10H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR