Anti-Epithelial Cell Surface Antigen Mouse Monoclonal Antibody [clone: 10B1416]

Supplier: United States Biological
USBIE3405-50EA 599 EUR
USBIE3405-50
Anti-Epithelial Cell Surface Antigen Mouse Monoclonal Antibody [clone: 10B1416]
Antibodies
Recommended dilution: Immunochemistry:1:200; Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: Epithelial Cell Surface Antigen
Clonality: monoclonal
Clone: 10B1416
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR