Anti-Germ Cell Nuclear Factor Rabbit polyclonal antibody

Supplier: United States Biological
USBIG3010-01EA 554 EUR
USBIG3010-01
Anti-Germ Cell Nuclear Factor Rabbit polyclonal antibody
Antibodies
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Germ Cell Nuclear Factor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR