Anti-Hepatocyte Cell Adhesion Molecule Sheep Polyclonal Antibody

Supplier: United States Biological
USBIH2004-50AEA 692 EUR
USBIH2004-50A
Anti-Hepatocyte Cell Adhesion Molecule Sheep Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Hepatocyte Cell Adhesion Molecule
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR