Anti-Mast Cell Chymase Sheep Polyclonal Antibody

Supplier: United States Biological
USBIM2414-01BEA 817 EUR
USBIM2414-01B
Anti-Mast Cell Chymase Sheep Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: Mast Cell Chymase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR