Anti-Mast Cell Protease I Goat Polyclonal Antibody

Supplier: United States Biological
USBIM2414-01JEA 531 EUR
USBIM2414-01J
Anti-Mast Cell Protease I Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: Mast Cell Protease I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR