Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: 12H35]

Supplier: United States Biological
USBIM2415-01DEA 989 EUR
USBIM2415-01D
Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: 12H35]
Antibodies
Recommended dilution: Immunochemistry: 1:10000

Type: Primary
Antigen: Mast Cell Tryptase
Clonality: monoclonal
Clone: 12H35
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat, Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR