Anti-MAP4K1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBIM3949-71AEA 587 EUR
USBIM3949-71A
Anti-MAP4K1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500

Type: Primary
Antigen: MAP4K1 (mitogen-activated protein kinase kinase kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR