Anti-NK Cell Activation Structures Mouse Monoclonal Antibody [clone: ANK61]

Supplier: United States Biological
USBIN2834-25EA 850 EUR
USBIN2834-25
Anti-NK Cell Activation Structures Mouse Monoclonal Antibody [clone: ANK61]
Antibodies
Type: Primary
Antigen: NK Cell Activation Structures
Clonality: monoclonal
Clone: ANK61
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR