Anti-Renal Cell Carcinoma Marker, Aminopeptidase A Mouse Monoclonal Antibody [clone: RC38]

Supplier: United States Biological
USBIR1450-03JEA 462 EUR
USBIR1450-03J
Anti-Renal Cell Carcinoma Marker, Aminopeptidase A Mouse Monoclonal Antibody [clone: RC38]
Antibodies
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Renal Cell Carcinoma Marker, Aminopeptidase A
Clonality: monoclonal
Clone: RC38
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR