Anti-Stem Cell Growth Factor Goat Polyclonal Antibody

Supplier: United States Biological
USBIS7973-24EA 783 EUR
USBIS7973-24
Anti-Stem Cell Growth Factor Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Stem Cell Growth Factor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR